Learn More
Alkali Scientific HPLC Chromatography Vial, 2mL Clear Glass vial with Writing Area, 10 Packs of 100, 1000/cs
SDP
Catalog No. 502335876
Encompass_Preferred
These HPLC Chromatography Vials are 2mL with a writing area, flat base, graduations, 9-425 open top ribbed screw cap with 9mm white PTFE/red silicone 1mm thick. The vial size is 12 x 32mm. They come in 10 packs of 100, 1000 per case. Application - Standard autosampler vial for HPLC and GC chromatography, sample vials can be used for high performance liquid chromatography and gas chromatography for reliable analysis results. A great deal of energy, money, and time can be spent on sample preparation. The last thing you need is for a sample vial to contribute to contamination resulting in complicated data interpretation or complete reanalysis. By using certified vials that are nearly contaminant free, you can reduce the risk significantly by using vial products made specifically for these applications.
Catalog No. 50-233-5876 Supplier Alkali Scientific Supplier No. CV5500
These HPLC Chromatography Vials are 2mL with a writing area, flat base, graduations, 9-425 open top ribbed screw cap with 9mm white PTFE/red silicone 1mm thick. The vial size is 12 x 32mm. They come in 10 packs of 100, 1000 per case. Application - Standard autosampler vial for HPLC and GC chromatography, sample vials can be used for high performance liquid chromatography and gas chromatography for reliable analysis results. A great deal of energy, money, and time can be spent on sample preparation. The last thing you need is for a sample vial to contribute to contamination resulting in complicated data interpretation or complete reanalysis. By using certified vials that are nearly contaminant free, you can reduce the risk significantly by using vial products made specifically for these applications.