Learn More
Alkali Scientific HPLC Chromatography Vial, 2mL AMBER Glass vial with Writing Area, Flat Base w/ graduations, 9-425 Open Top Ribbed Screw Cap with 9mm White PTFE/Red Silicone Septa 1mm Thick, Vial size: 12 x 32 mm, 1000/case
SDP
Catalog No. 503027197
Encompass_Preferred
HPLC Chromatography VialA great deal of energy, money, and time can be spent on sample preparation. The last thing you need is for a sample vial to contribute to contamination resulting in complicated data interpretation or complete reanalysis. By using certified vials that are nearly contaminant free, you can reduce the risk significantly by using vial products made specifically for these applications. Vial Material : Amber Type 1, Class A Borosilicate Glass Easy to Use : 9-425 screw top vials is 40% larger opening than standard narrow opening vials, 2ml HPLC Vials with Unique Thread Design Ensures Consistent Seal and Can Be Reused, Not Volatile Vial insert Dimensions: 12 x 32 mml Ribbed Screw Cap with PTFE/Silicone Septa : 2ml clear autosampler vial included blue ABS Screw Cap & Septa (Red PTFE/White Silicone Liner) This PTFE/Silicone septa with PTFE chemically inert before being penetrated and strongest repetitive sealing ability-recommended for multiple penetration sampling
Catalog No. 50-302-7197 Supplier Alkali Scientific Supplier No. CV5600CS
HPLC Chromatography VialA great deal of energy, money, and time can be spent on sample preparation. The last thing you need is for a sample vial to contribute to contamination resulting in complicated data interpretation or complete reanalysis. By using certified vials that are nearly contaminant free, you can reduce the risk significantly by using vial products made specifically for these applications. Vial Material : Amber Type 1, Class A Borosilicate Glass Easy to Use : 9-425 screw top vials is 40% larger opening than standard narrow opening vials, 2ml HPLC Vials with Unique Thread Design Ensures Consistent Seal and Can Be Reused, Not Volatile Vial insert Dimensions: 12 x 32 mml Ribbed Screw Cap with PTFE/Silicone Septa : 2ml clear autosampler vial included blue ABS Screw Cap & Septa (Red PTFE/White Silicone Liner) This PTFE/Silicone septa with PTFE chemically inert before being penetrated and strongest repetitive sealing ability-recommended for multiple penetration sampling