Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 150 x 4.6mm

Catalog No. 50156048 Shop All Phenomenex Products
Encompass

Venusil 5m XBP-C18(2) 100A VX951505-2

Catalog No. 50-156-048 Supplier Phenomenex Inc Supplier No. VX9515052
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Venusil 5m XBP-C18(2) 100A VX951505-2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.