Learn More
Microsolv Technology Corporation HPLC Column, Amide, 4um, 4.6mm ID x 100mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 4003610P
Encompass_Preferred
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This size column is often used when shorter run times are needed but 50mm long columns do not provide enough resolution.
Catalog No. 50-227-0759