Learn More
Microsolv Technology Corporation HPLC Column, Amide, 4um, 4.6mm ID x 150mm Length, 100A. 1 EA.
Catalog No. 502270761
Encompass_Preferred
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column is one of the more popular column sizes and often chosen for use with standard LC systems that have ELSD or Corona CAD detectors or are working with UV absorbing nitrogen containing compounds.
This item is not returnable. View return policy
Catalog No. 50-227-0761 Supplier Microsolv Technology Corporation Supplier No. 4003615P
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column is one of the more popular column sizes and often chosen for use with standard LC systems that have ELSD or Corona CAD detectors or are working with UV absorbing nitrogen containing compounds.