Learn More
Microsolv Technology Corporation HPLC Column, Amide, 4um, 4.6mm ID x 250mm Length, 100A. 1 EA.
Catalog No. 502270763
Encompass_Preferred
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column was one of the more popular column sizes and often chosen for use with standard LC systems that have ELSD or Corona CAD detectors or are working with UV absorbing nitrogen containing compounds. Chosen when more separation between closely related compounds or longer retention times is needed .
This item is not returnable. View return policy
Catalog No. 50-227-0763 Supplier Microsolv Technology Corporation Supplier No. 4003625P
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column was one of the more popular column sizes and often chosen for use with standard LC systems that have ELSD or Corona CAD detectors or are working with UV absorbing nitrogen containing compounds. Chosen when more separation between closely related compounds or longer retention times is needed .