Learn More
Microsolv Technology Corporation Prep HPLC Column, Amide, 4um, 100A. 21.2mm ID x 150mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 40036P21150
Encompass_Preferred
This Cogent Amide Preparative, HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for Purification or "Prep" of Sugars and other Analytes with amine groups when a large volume of sample is desired to be collected. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0774
Add to Cart
Request a Quote