Learn More
Microsolv Technology Corporation HPLC Column, Amide, 4um, 2.1mm ID x 150mm Length, 100A. 1 EA.
Catalog No. 502270762
Encompass_Preferred
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column is often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-227-0762 Supplier Microsolv Technology Corporation Supplier No. 4003615P2
This Cogent Amide HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of Sugars and other Analytes with amine groups, This column is often chosen for LCMS when increased resolution is desired.