Learn More
Microsolv Technology Corporation HPLC Column, Bidentate C18 2.o, 2.2um, 2.1mm ID x 50mm Length, 120A. 1 EA.
Supplier:  Microsolv Technology Corporation 4021805P2
Encompass_Preferred
This Cogent Bidentate C18 2.o HPLC Column is made with Cogent TYPE-C™ Silica , and this smaller particle, silica hydride based stationary phase provides Increased Efficiency, Excellent Selectivity and Faster Equilibration. Considered "Near UHPLC", this column can be used on any HPLC Instrument and still deliver improved efficiency. Chosen for analysis of hydrophobic compounds that are difficult to retain in Aqueous Mobile Phases, including very non-polar compounds. This column is considered a Mass Spec Friendly Column. This short and narrow bore column provides short run times but requires a pump that can precisely deliver, and a flow cell that made for, reduced flow rates with lower volumes. This column size is excellent, and a favorite for LCMS.
Catalog No. 50-227-0830