Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, Bidentate C8, 4um, 100A. 10mm ID x 100mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 40008SP100
Encompass_Preferred
This Cogent Bidentate C8 "Semi-Preparative", HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Very Robust Column Lifetimes. Chosen for Purification or "Prep" of Hydrophobic Molecules, Metabolites, and other Analytes with amine groups when a large volume of sample is desired to be collected and C18 is too retentive. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0697