Learn More
Microsolv Technology Corporation HPLC Column, Diamond Hydride, 4um, 3.0mm ID x 150mm Length, 100A. 1 EA.
Catalog No. 502270928
Encompass_Preferred
This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. This size column provides similar run times as a 4.6mm ID column, but uses much less mobile phase, with lower flow rates. No special equipment is needed for this column to save solvent use
This item is not returnable. View return policy
Catalog No. 50-227-0928 Supplier Microsolv Technology Corporation Supplier No. 7000015P3
This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. This size column provides similar run times as a 4.6mm ID column, but uses much less mobile phase, with lower flow rates. No special equipment is needed for this column to save solvent use