Learn More
Microsolv Technology Corporation HPLC Column, Diamond Hydride, 4um, 4.6mm ID x 250mm Length, 100A. 1 EA.
Catalog No. 502270929
Encompass_Preferred
This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Chosen when more separation between closely related compounds or longer retention times is needed.
This item is not returnable. View return policy
Catalog No. 50-227-0929 Supplier Microsolv Technology Corporation Supplier No. 7000025P
This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Chosen when more separation between closely related compounds or longer retention times is needed.