Learn More
Microsolv Technology Corporation HPLC Column, Diol, 4um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Catalog No. 502270784
Encompass_Preferred
This Cogent Diol HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Used in Reversed Phase, Normal Phase or Aqueous Normal Phase (ANP). This size column provides good resolution and short run times and is a Very Popular Column used in High Through-put Screening.
This item is not returnable. View return policy
Catalog No. 50-227-0784 Supplier Microsolv Technology Corporation Supplier No. 4006005P
This Cogent Diol HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Used in Reversed Phase, Normal Phase or Aqueous Normal Phase (ANP). This size column provides good resolution and short run times and is a Very Popular Column used in High Through-put Screening.