Learn More
Microsolv Technology Corporation HPLC Column, Diol, 4um, 4.6mm ID x 100mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 4006010P
Encompass_Preferred
This Cogent Diol HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Used in Reversed Phase, Normal Phase or Aqueous Normal Phase (ANP).
Catalog No. 50-227-0786