Learn More
Microsolv Technology Corporation HPLC Column, Diol, 4um, 2.1mm ID x 100mm Length, 100A. 1 EA.
Catalog No. 502270787
Encompass_Preferred
This Cogent Diol HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Used in Reversed Phase, Normal Phase or Aqueous Normal Phase (ANP). This size column provides good resolution and good run times and is a Very Popular Column used with LCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0787 Supplier Microsolv Technology Corporation Supplier No. 4006010P2
Add to Cart
Add to Cart
This Cogent Diol HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. Used in Reversed Phase, Normal Phase or Aqueous Normal Phase (ANP). This size column provides good resolution and good run times and is a Very Popular Column used with LCMS.