Learn More
Microsolv Technology Corporation HPLC Column, HPS Amino, 5um, 4.6mm ID x 100mm Length, 120A. 1 EA
Catalog No. 502270982
Encompass_Preferred
This Cogent HPS Amino™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. Not end capped, it is manufactured using tri-functional bonding of a 3 carbon silane with a terminal amino group to our unique High Purity Silica. This Column is ideal as an Alternate in any USP Method that uses the L8 Classification or for new methods. With a 5 µm Particle, this Column works well with higher flow rates and lower back pressures compared with smaller particles. Used for polar compounds with a negative charge such as Sugars and compounds with Carboxylic Acid Groups in Normal Phase or HILIC modes. Compare to Inertsil ™ Amino and Luna ™ Amino Columns.
This item is not returnable. View return policy
Catalog No. 50-227-0982 Supplier Microsolv Technology Corporation Supplier No. 7504010P
This Cogent HPS Amino™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. Not end capped, it is manufactured using tri-functional bonding of a 3 carbon silane with a terminal amino group to our unique High Purity Silica. This Column is ideal as an Alternate in any USP Method that uses the L8 Classification or for new methods. With a 5 µm Particle, this Column works well with higher flow rates and lower back pressures compared with smaller particles. Used for polar compounds with a negative charge such as Sugars and compounds with Carboxylic Acid Groups in Normal Phase or HILIC modes. Compare to Inertsil ™ Amino and Luna ™ Amino Columns.