Learn More
Microsolv Technology Corporation HPLC Column, HPS Phenyl, 5um, 4.6mm ID x 100mm Length, 120A. 1 EA.
Catalog No. 502271002
Encompass_Preferred
This Cogent HPS Phenyl™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is fully end capped and is manufactured with a mono functional, 4 carbon ligand that has an attached Phenyl group and it is bonded to the highest purity, 120A silica available making it ideal as an Alternate in USP Methods that use the L11 Classification. With a 5µm Particle, this Column works well with higher flow rates producing lower back pressures compared to columns with smaller particles. It is often used to separate Isomers (positional), Tocopherols, Flavonoids, Polynuclear Aromatics, Nitro-Aromatic Compounds, Active Pharmaceutical Ingredients (APIs), Natural Products and Related Compounds in Reversed Phase HPLC. Compare this column's performance to Luna ™ or Inertsil ™ columns.
This item is not returnable. View return policy
Catalog No. 50-227-1002 Supplier Microsolv Technology Corporation Supplier No. 7502010P
This Cogent HPS Phenyl™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is fully end capped and is manufactured with a mono functional, 4 carbon ligand that has an attached Phenyl group and it is bonded to the highest purity, 120A silica available making it ideal as an Alternate in USP Methods that use the L11 Classification. With a 5µm Particle, this Column works well with higher flow rates producing lower back pressures compared to columns with smaller particles. It is often used to separate Isomers (positional), Tocopherols, Flavonoids, Polynuclear Aromatics, Nitro-Aromatic Compounds, Active Pharmaceutical Ingredients (APIs), Natural Products and Related Compounds in Reversed Phase HPLC. Compare this column's performance to Luna ™ or Inertsil ™ columns.