Learn More
Microsolv Technology Corporation HPLC Column, HPS Silica, 5um, 2.1mm ID x 50mm Length, 120A. 1 EA.
Catalog No. 502271007
Encompass_Preferred
This Cogent HPS Silica™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. This proprietary Silica is not modified in any way and is ideal as an Alternate in any USP Method that uses the L3 Classification or for new methods. With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles and is often used for hydrophilic or polar compounds in Normal Phase HPLC. Compare its performance to Inertsil and Luna products.
This item is not returnable. View return policy
Catalog No. 50-227-1007 Supplier Microsolv Technology Corporation Supplier No. 7500105P2
This Cogent HPS Silica™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. This proprietary Silica is not modified in any way and is ideal as an Alternate in any USP Method that uses the L3 Classification or for new methods. With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles and is often used for hydrophilic or polar compounds in Normal Phase HPLC. Compare its performance to Inertsil and Luna products.