Learn More
Microsolv Technology Corporation HPLC Column, IEX WAX, 5um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 6759005P
Encompass_Preferred
This Cogent Ion Exchange Column is a Weak Anion Exchanger (WAX has a positively charged surface ) Stationary Phase that has been a premium value and used by HPLC laboratories for many years. It is manufactured with Diethylaminopropyl ligands that have a functional charged range between pH 5 and 8. We use only the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L9 Classification or for acids analysis or purification. With a 5µm Particle, this column works well and produces excellent data for challenging IEX applications that include Anions (negatively charge compounds).
Catalog No. 50-226-9985