Learn More
Microsolv Technology Corporation HPLC Column, IEX WCX, 10um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Catalog No. 502269989
Encompass_Preferred
This Cogent Ion Exchange Column is a Weak Cation Exchanger (WCX has a negatively charged surface) Stationary Phase that has been a premium value and used by HPLC laboratories for many years. It is manufactured with ligands that include a carboxylic acid producing a functional charged range between pH 2.5 and 7. We use only the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L9 Classification or for "base" analysis or purification. With a 10µm Particle, this column works well with higher flow rates and lower back pressures for challenging IEX applications that include Cations (positively charged compounds).
This item is not returnable. View return policy
Catalog No. 50-226-9989 Supplier Microsolv Technology Corporation Supplier No. 6709105P
This Cogent Ion Exchange Column is a Weak Cation Exchanger (WCX has a negatively charged surface) Stationary Phase that has been a premium value and used by HPLC laboratories for many years. It is manufactured with ligands that include a carboxylic acid producing a functional charged range between pH 2.5 and 7. We use only the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L9 Classification or for "base" analysis or purification. With a 10µm Particle, this column works well with higher flow rates and lower back pressures for challenging IEX applications that include Cations (positively charged compounds).