Learn More
Mac-Mod Avantor Partisil C8, 85 A, 10 um, 4.6 x 250 mm HPLC Column
SDP
Supplier:  Mac-Mod 4229001
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-761