Learn More
Mac-Mod Avantor Partisil ODS, 85 , 10 m, 4.6 x 250 mm HPLC Column (WCS hardware)
SDP
Catalog No. 50304755
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
This item is not returnable. View return policy
Catalog No. 50-304-755 Supplier Mac-Mod Supplier No. 4223001
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.