Learn More
Mac-Mod Avantor Partisil PAC, 85 A, 5 um, 4.6 x 250 mm HPLC Column (WCS hardware)
SDP
Catalog No. 50304751
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-751 Supplier Mac-Mod Supplier No. 4235001
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.