Learn More
Mac-Mod Avantor Partisil SAX, 85 A, 5 um, 4.6 x 100 mm HPLC Column (WCS hardware - RAC-II)
SDP
Supplier:  Mac-Mod 4222227
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-747