Learn More
Mac-Mod Avantor Partisil SCX, 85 A, 5 um, 4.6 x 250 mm HPLC Column (WVS hardware)
SDP
Catalog No. 50304790
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-790 Supplier Mac-Mod Supplier No. 46811507
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.