Learn More
Mac-Mod Avantor Partisil Silica, 85 A, 10 um, 9.4 x 250 mm HPLC Column (Magnum-9 hardware)
SDP
Catalog No. 50304763
Encompass
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-763 Supplier Mac-Mod Supplier No. 4230120
Avantor Partisil is a high-purity irregular silica gel packed in HPLC columns. Avantor Partisil columns provide reproducible results, column to column, lot to lot.