Learn More
Mac-Mod Avantor Partisphere SCX, 120 A, 5 um, 4.6 x 125 mm HPLC Column (WVS hardware)
SDP
Supplier:  Mac-Mod 46210507
Encompass
Avantor Partisphere is a high-purity gel packed in HPLC Columns. Avantor Partisphere columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-304-778