Learn More
Microsolv Technology Corporation HPLC Column, Phenyl Hydride 2.o, 2.2um, 2.1mm ID x 150mm Length, 120A, High Pressure. 1 EA.
Supplier:  Microsolv Technology Corporation 6922015P2
Encompass_Preferred
This Cogent Phenyl Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for purification of compounds with aromatic rings that are difficult to retain or separate in Reversed Phase, including aromatic amines, small peptides and azides. This very popular column can be used for LCMS when increased resolution is desired.
Catalog No. 50-227-0914