Learn More
Microsolv Technology Corporation HPLC Column, Phenyl Hydride, 4um, 4.6mm ID x 150mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 6902015P
Encompass_Preferred
This Cogent Phenyl Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of compounds with aromatic rings that are difficult to retain or separate in Reversed Phase, including aromatic amines, small peptides and azides. This column is one of our more popular column sizes and often chosen for use in Aqueous Normal Phase mode for fast analysis of small, very polar compounds in HPLC using UV detection.
Catalog No. 50-227-0863