Learn More
Microsolv Technology Corporation HPLC Column, Phenyl Hydride, 4um, 4.6mm ID x 50mm Length, 100A, 1 EA.
Supplier:  Microsolv Technology Corporation 6902005P
Encompass_Preferred
This Cogent Phenyl Hydride HPLC Column is made with Cogent TYPE-C™ Silica , and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes. Chosen for analysis of compounds with aromatic rings that are difficult to retain or separate in Reversed Phase, including aromatic amines, small peptides and azides. This size column provides good resolution with short run times and is a very popular column used in High Through-put Screening.
Catalog No. 50-227-0859