Learn More
Microsolv Technology Corporation HPLC Column, RP C18, Wide Pore, 5um, 4.6mm ID x 50mm Length, 300A. 1 EA.
Catalog No. 502269852
Encompass_Preferred
This Cogent RP C18™ Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available.With a 5 µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. Used for large hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is also excellent for long chain fatty acids, some proteins and peptides. This size column provides good resolution and short run times and is Very Popular Column used in High Through-put Screening.
This item is not returnable. View return policy
Catalog No. 50-226-9852 Supplier Microsolv Technology Corporation Supplier No. 6851805P3M
This Cogent RP C18™ Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available.With a 5 µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. Used for large hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is also excellent for long chain fatty acids, some proteins and peptides. This size column provides good resolution and short run times and is Very Popular Column used in High Through-put Screening.