Learn More
Microsolv Technology Corporation HPLC Column, RP C18, 5um, 2.1mm ID x 150mm Length, 100A. 1 EA.
Catalog No. 502269858
Encompass_Preferred
This Cogent RP C18™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L1 Classification.With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles. Used for hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is excellent for long chain fatty acids. Expect longer retention than with a C8 column. This column is often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-226-9858 Supplier Microsolv Technology Corporation Supplier No. 6851815P2
This Cogent RP C18™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available, making this column ideal as an Alternate in USP Methods that use the L1 Classification.With a 5 µm Particle, this column works well with higher flow rates and lower back pressures compared with smaller particles. Used for hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is excellent for long chain fatty acids. Expect longer retention than with a C8 column. This column is often chosen for LCMS when increased resolution is desired.