Learn More
Microsolv Technology Corporation HPLC Column, RP C18, Wide Pore, 5um, 4.6mm ID x 150mm Length, 300A. 1 EA.
Catalog No. 502269860
Encompass_Preferred
This Cogent RP C18™ Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available.With a 5 µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. Used for large hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is also excellent for long chain fatty acids, some proteins and peptides. This column is often chosen for use macro-molecules such as proteins and polysaccharides in HPLC using UV detection or in LCMS with a flow splitter.
This item is not returnable. View return policy
Catalog No. 50-226-9860 Supplier Microsolv Technology Corporation Supplier No. 6851815P3M
This Cogent RP C18™ Wide Pore Stationary Phase has been a premium value and used by HPLC laboratories for many years. It is double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available.With a 5 µm Particle and 300A Pore Size, this column works well with higher flow rates and lower back pressures. Used for large hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is also excellent for long chain fatty acids, some proteins and peptides. This column is often chosen for use macro-molecules such as proteins and polysaccharides in HPLC using UV detection or in LCMS with a flow splitter.