Learn More
Microsolv Technology Corporation HPLC Column, RP C18 PE, 5um, 4.6mm ID x 100mm Length, 100A. 1 EA.
Catalog No. 502270231
Encompass_Preferred
This Cogent RP C18 PE™ Column is a premium value and used in HPLC with polar and non polar compounds. Double end capped (fully), it is bonded with an 18 carbon aliphatic hydrocarbon, with an Amide group embedded in the middle of the ligand, using the highest purity silica available. This Column is an ideal Alternate for your USP Methods that uses the L1 Classification and 100% Water as the Mobile Phase for hydrophobic samples. Very rugged, and often called "Polar Embedded", it can be used in typical Reversed Phase or HILIC modes. Used for hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is excellent for long chain fatty acids.
This item is not returnable. View return policy
Catalog No. 50-227-0231 Supplier Microsolv Technology Corporation Supplier No. 8451810P
This Cogent RP C18 PE™ Column is a premium value and used in HPLC with polar and non polar compounds. Double end capped (fully), it is bonded with an 18 carbon aliphatic hydrocarbon, with an Amide group embedded in the middle of the ligand, using the highest purity silica available. This Column is an ideal Alternate for your USP Methods that uses the L1 Classification and 100% Water as the Mobile Phase for hydrophobic samples. Very rugged, and often called "Polar Embedded", it can be used in typical Reversed Phase or HILIC modes. Used for hydrophobic or neutral compounds in Reversed Phase, this Octadecyl Phase is excellent for long chain fatty acids.