Learn More
Sepax Technologies Inc HPLC Column, Sepax, SRT-LVL SEC-2000, 5µm, 2000 Å, 7.8 x 300 mm
SDP
Catalog No. NC4020363
Encompass
HPLC Column, Sepax, SRT-LVL SEC-2000, 5µm, 2000 Å, 7.8 x 300 mm
Catalog No. NC4020363 Supplier Sepax Technologies Inc Supplier No. 2199807830
HPLC Column, Sepax, SRT-LVL SEC-2000, 5µm, 2000 Å, 7.8 x 300 mm