Learn More
Microsolv Technology Corporation HPLC Column, Silica-C, 4um, 4.6mm ID x 100mm Length, 100A. 1 EA.
Supplier:  Microsolv Technology Corporation 4000010P
Encompass_Preferred
The Silica-C™ HPLC Columns are made with Cogent TYPE-C™ Silica, and provide unique Selectivity due to its surface chemistry (especially in Normal Phase HPLC) compared to standard silica, This Silica Hydride Phase is not modified and therefore has no end-capping or organic ligands. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster Equilibration than other Silica Columns, no need to control moisture in Mobile Phase Solvents and robust column lifetimes. This size column is often used when shorter run times are needed but 50mm long columns do not provide enough resolution.
Catalog No. 50-227-0631