Learn More
Microsolv Technology Corporation HPLC Column, Silica-C 2.o, 2.2um, 2.1mm ID x 100mm Length, 120A. 1 EA..
Catalog No. 502270819
Encompass_Preferred
This Cogent Silica-C 2.o™ “near UHPLC”, HPLC Column is made with 2.2um, 120A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes This column is one of the more popular 2.o column sizes and often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-227-0819 Supplier Microsolv Technology Corporation Supplier No. 4020010P2
This Cogent Silica-C 2.o™ “near UHPLC”, HPLC Column is made with 2.2um, 120A, Cogent TYPE-C™ Silica , and this silica hydride based stationary phase. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes This column is one of the more popular 2.o column sizes and often chosen for LCMS when increased resolution is desired.