Learn More
Microsolv Technology Corporation HPLC Column, UDA (wcx) 2.o, 2.2um, 2.1mm ID x 50mm Length, 120A, High. 1 EA.
Supplier:  Microsolv Technology Corporation 4023105P2
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group and is considered a “near UHPLC” Column with 2.2um, 120A, silica hydride as the base silica.Use in Reversed Phase (RP), Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes Use Minimal Salt to Elute in ANP This short and narrow bore column provides short run times but requires a pump that can precisely deliver, and a flow cell that made for, reduced flow rates with lower volumes.
Catalog No. 50-227-0836