Learn More
Microsolv Technology Corporation Prep HPLC Column, Silica-C, 10um, 100A. 50mm ID x 50mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 40000P5050
Encompass_Preferred
This Cogent Silica Hydride Preparative, HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica, and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes.Chosen for Purification or "Prep" of Peptides, Metabolites, Polar Compounds and other Analytes with amine groups when a large volume of sample is desired to be collected. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0656
Request a Quote
Please call customer service for current pricing. 1-800-766-7000