Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, Silica-C, 4um, 100A. 10mm ID x 250mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 40000SP250
Encompass_Preferred
This Cogent Silica-C™ "Semi" Preparative, HPLC Column is made with 4um, 100A, Cogent TYPE-C™ Silica, and this silica hydride based stationary phase provides excellent yields and turn around times. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster Equilibration than other silica columns, no need to control moisture in Mobile Phase Solvents, robust column lifetimes. Use Minimal Salt to Elute in ANP. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-0659