Learn More
Microsolv Technology Corporation HPLC Column, Silica-C, 4um, 2.1mm ID x 50mm Length, 100A. 1 EA.
Catalog No. 502270630
Encompass_Preferred
The Silica-C™ HPLC Columns are made with Cogent TYPE-C™ Silica, and provide unique Selectivity due to its surface chemistry (especially in Normal Phase HPLC) compared to standard silica, This Silica Hydride Phase is not modified and therefore has no end-capping or organic ligands. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster Equilibration than other Silica Columns, no need to control moisture in Mobile Phase Solvents and robust column lifetimes. This column size is excellent, and a favorite for LCMS.
This item is not returnable. View return policy
Catalog No. 50-227-0630 Supplier Microsolv Technology Corporation Supplier No. 4000005P2
Add to Cart
Add to Cart
The Silica-C™ HPLC Columns are made with Cogent TYPE-C™ Silica, and provide unique Selectivity due to its surface chemistry (especially in Normal Phase HPLC) compared to standard silica, This Silica Hydride Phase is not modified and therefore has no end-capping or organic ligands. Chosen for analyzing compounds that are not soluble in Water including but not limited to: lipids, peptides, proteins, oligonucleotides and other compounds that require non polar solvents. Faster Equilibration than other Silica Columns, no need to control moisture in Mobile Phase Solvents and robust column lifetimes. This column size is excellent, and a favorite for LCMS.