Learn More
Microsolv Technology Corporation HPLC Column, UDA (wcx), 4um, 4.6mm ID x 100mm Length, 100A. 1 EA..
Catalog No. 502270731
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP) , Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). The hydrophobic stationary phase has direct silicon-carbon bonds which makes the column very robust and can be used for hydrophilic or hydrophobic compounds, if desired. Also can be used at very low pH (below 2) without damage. This size column is often used when shorter run times are needed but 50mm long columns do not provide enough resolution.
This item is not returnable. View return policy
Catalog No. 50-227-0731 Supplier Microsolv Technology Corporation Supplier No. 4003110P
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP) , Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). The hydrophobic stationary phase has direct silicon-carbon bonds which makes the column very robust and can be used for hydrophilic or hydrophobic compounds, if desired. Also can be used at very low pH (below 2) without damage. This size column is often used when shorter run times are needed but 50mm long columns do not provide enough resolution.