Learn More
Microsolv Technology Corporation HPLC Column, UDA (wcx), 4um, 2.1mm ID x 150mm Length, 100A. 1 EA.
Catalog No. 502270734
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP) , Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). The hydrophobic stationary phase has direct silicon-carbon bonds which makes the column very robust and can be used for hydrophilic or hydrophobic compounds, if desired. Also can be used at very low pH (below 2) without damage. This column is often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-227-0734 Supplier Microsolv Technology Corporation Supplier No. 4003115P2
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP) , Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). The hydrophobic stationary phase has direct silicon-carbon bonds which makes the column very robust and can be used for hydrophilic or hydrophobic compounds, if desired. Also can be used at very low pH (below 2) without damage. This column is often chosen for LCMS when increased resolution is desired.