Learn More
Microsolv Technology Corporation Prep HPLC Column, UDA (wcx), 4um, 100A. 21.2mm ID x 150mm Length, Preparative. 1 EA.
Supplier:  Microsolv Technology Corporation 40031P21150
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group. This silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Extremely Robust Column Lifetimes.Use in Reversed Phase (RP), Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes Use Minimal Salt to Elute in ANP A specialized HPLC System is required for columns this size .
Catalog No. 50-227-0746
Add to Cart
Request a Quote