Learn More
Microsolv Technology Corporation HPLC Column, UDA (wcx) 2.o, 2.2um, 2.1mm ID x 150mm Length, 120A. 1 EA.
Catalog No. 502270838
Encompass_Preferred
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group and is considered a “near UHPLC” Column with 2.2um, 120A, silica hydride as the base silica.Use in Reversed Phase (RP), Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes Use Minimal Salt to Elute in ANP This column is often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-227-0838 Supplier Microsolv Technology Corporation Supplier No. 4023115P2
This Cogent UDA™ (wcx) HPLC Column is made with Cogent TYPE-C™ Silica , and has an 11 carbon chain with a terminal carboxylic acid group and is considered a “near UHPLC” Column with 2.2um, 120A, silica hydride as the base silica.Use in Reversed Phase (RP), Aqueous Normal Phase (ANP) or as a Weak Cation Exchange Column (wcx at pH 6 or higher). Faster equilibration than other Silica columns, no need to control moisture in mobile phase solvents and robust column lifetimes Use Minimal Salt to Elute in ANP This column is often chosen for LCMS when increased resolution is desired.