Learn More
Microsolv Technology Corporation HPLC Column, UDC Cholesterol, 4um, 2.1mm ID x 100mm Length, 100A. 1 EA.
Catalog No. 502270889
Encompass_Preferred
This UDC Cholesterol™ HPLC is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11 carbon chain with a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than other Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity This column is very popular and often chosen for LCMS when increased resolution is desired.
This item is not returnable. View return policy
Catalog No. 50-227-0889 Supplier Microsolv Technology Corporation Supplier No. 6906910P2
This UDC Cholesterol™ HPLC is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11 carbon chain with a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than other Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity This column is very popular and often chosen for LCMS when increased resolution is desired.