Learn More
Microsolv Technology Corporation HPLC Column, UDC Cholesterol, 4um, 4.6mm ID x 50mm Length, 100A. 1 EA.
Catalog No. 502270887
Encompass_Preferred
This UDC Cholesterol™ Column is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11 carbon chain that has a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity This size column provides good resolution and short run times and is used in High Through-put Screening.
This item is not returnable. View return policy
Catalog No. 50-227-0887 Supplier Microsolv Technology Corporation Supplier No. 6906905P
Add to Cart
Add to Cart
This UDC Cholesterol™ Column is made with Cogent TYPE-C™ Silica , and provides unique Selectivity due to different surface chemistry, compared to typical Reversed Phase Columns. This Silica Hydride Phase is modified with an 11 carbon chain that has a cholesterol molecule attached at the terminus. This gives this column the ability to form “slots” and separate some compounds based on shape. Also, due to the direct silicon carbon bonds, this column has no need for end-capping. Use in Reversed Phase, Normal Phase and Aqueous Normal Phase (ANP). Chosen for analyzing steroidal and other hard to retain compounds including some lipids, peptides, proteins & oligonucleotides. Faster Equilibration than Reversed Phase or HILIC Columns Very Hydrophobic Robust Column Lifetimes Temperature and Solvent Changes Selectivity This size column provides good resolution and short run times and is used in High Through-put Screening.