Learn More
Trajan Scientific HPLC Ferrule PEEK Nut 2/PK
SDP
Catalog No. 5000494264
Encompass_Preferred
Stainless steel hexnuts and PEEK ferrules 2 hexnuts and 2 ferrules
Catalog No. 50-004-94264 Supplier Trajan Scientific Supplier No. 1021003
Stainless steel hexnuts and PEEK ferrules 2 hexnuts and 2 ferrules