Learn More
Trajan Scientific HPLC Fitting PEEKLok Medical 10/PK
SDP
Catalog No. 5000494053
Encompass_Preferred
PEEKLOK 1/16 10-32 FITTINGS PK10
Catalog No. 50-004-94053 Supplier Trajan Scientific Supplier No. 225051
PEEKLOK 1/16 10-32 FITTINGS PK10