Learn More
ANALYTICS SHOP USA LP HPLC GUARD CARTRIDGE ZIRCHROM
SDP
Catalog No. NC3891574
Encompass
HPLC Guard Cartridge ZirChrom-EZ, G20, for 2.1 mm Column, Holder ZC-852-00 needed, 3 pc/PAK
Catalog No. NC3891574 Supplier ANALYTICS SHOP USA LP Supplier No. ZCEZ01G20
HPLC Guard Cartridge ZirChrom-EZ, G20, for 2.1 mm Column, Holder ZC-852-00 needed, 3 pc/PAK